1-Bromo-2-ethylbutane ≥95%

Supplier: Matrix Scientific

Synonyms: 3-(Bromomethyl)pentane

084166-1G 084166-10G
102841-694EA 272.31 USD
102841-694 102841-692
1-Bromo-2-ethylbutane ≥95%
1-Bromo-2-ethylbutane

Hazard: Irritant

Formula: C₆H₁₃Br
MW: 165.07 g/mol
MDL Number: MFCD00000219
CAS Number: 3814-34-4
UN: 1993
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR