3-Bromo-1-propanol ≥97%

Supplier: Matrix Scientific

Synonyms: 3-Bromopropan-1-ol1-Bromo-3-hydroxypropane

084161-100G
102841-676EA 136.92 USD
102841-676
3-Bromo-1-propanol ≥97%
3-Bromo-1-propanol

Hazard: Irritant

Formula: BrH₂CCH₂CH₂OH
MW: 138.99 g/mol
Boiling Pt: 71…73 °C (11 mmHg)
Density: 1.59
Flash Pt: 93 °C
MDL Number: MFCD00002942
CAS Number: 627-18-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR