3-Bromo-7-methylimidazo[1,2-a]pyridine ≥97%
Supplier: Matrix Scientific
084141-1G
102841-614EA
529
USD
102841-614
3-Bromo-7-methylimidazo[1,2-a]pyridine ≥97%
3-Bromo-7-methylimidazo[1,2-a]pyridine
Hazard: Irritant
Formula:
C₈H₇BrN₂ MW: 211.0600 g/mol |
MDL Number:
MFCD08056208 CAS Number: 56051-32-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...