8-Bromotheophylline ≥97%

Supplier: Matrix Scientific

084106-1G
102841-532EA 149.5 USD
102841-532
8-Bromotheophylline ≥97%
8-Bromotheophylline

Hazard: Irritant

Formula: C₇H₇BrN₄O₂
MDL Number: MFCD00022664
CAS Number: 10381-75-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR