7-Chloro-5-nitro-1H-indazole ≥97%

Supplier: Matrix Scientific

084092-1G
102841-504EA 949.23 USD
102841-504
7-Chloro-5-nitro-1H-indazole ≥97%
7-Chloro-5-nitro-1H-indazole

Hazard: Irritant

Formula: C₇H₄ClN₃O₂
CAS Number: 74209-22-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR