3-Iodo-5-nitro-1H-indazole ≥97%

Supplier: Matrix Scientific

084091-1G
102841-502EA 540.5 USD
102841-502
3-Iodo-5-nitro-1H-indazole ≥97%
3-Iodo-5-nitro-1H-indazole

Hazard: Irritant

Formula: C₇H₄IN₃O₂
MW: 289.0300 g/mol
MDL Number: MFCD07781630
CAS Number: 70315-69-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR