Monoethyl-4-oxoheptanedioate ≥97%
Supplier: Matrix Scientific
Synonyms:
4-Oxoheptanedioic Acid Monoethyl Ester
, 7-Ethoxy-4,7-dioxoheptanoic acid
083058-1G
083058-10G
102838-810EA
241.54
USD
102838-810
102838-812
Monoethyl-4-oxoheptanedioate ≥97%
Monoethyl-4-oxoheptanedioate
Purity: 97%
CAS: 1506-55-4
MDL Number: MFCD00059093
Molecular Formula: C9H14O5
Molecular Weight: 202,21
Hazard: Irritant
Formula:
C₉H₁₄O₅ MW: 202.21 g/mol |
MDL Number:
MFCD00059093 CAS Number: 1506-55-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...