cis-4,7,10,13,16,19-Docosahexaenoic acid, Technical Grade
Supplier: Matrix Scientific
Synonyms:
Docosa-4Z,7Z,10Z,13Z,16Z,19Z-Hexaenoic Acid
, DHA
083042-1G
102838-752EA
769.23
USD
102838-752
cis-4,7,10,13,16,19-Docosahexaenoic acid, Technical Grade
cis-4,7,10,13,16,19-Docosahexaenoic acid
CAS: 6217-54-5
MDL Number: MFCD13195609
Molecular Formula: C22H32O2
Molecular Weight: 328,5
Hazard: Irritant
Formula:
C₂₂H₃₂O₂ MW: 328.50 g/mol |
MDL Number:
MFCD00065722 CAS Number: 6217-54-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...