ɑ-Methyllevulinic acid ≥97%

Supplier: Matrix Scientific

Synonyms: 2-Methyl-4-oxopentanoic Acid

083040-1G
102838-744EA 298.46 USD
102838-744
ɑ-Methyllevulinic acid ≥97%
alpha-Methyllevulinic acid

Purity: 97%
CAS: 6641-83-4
MDL Number: MFCD00143584
Molecular Formula: C6H10O3
Molecular Weight: 130,14
Hazard: Irritant

Formula: C₆H₁₀O₃
MW: 130.14 g/mol
MDL Number: MFCD00143584
CAS Number: 6641-83-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR