4-Acetamidobutyric acid ≥97%
Supplier: Matrix Scientific
Synonyms:
4-Acetamidobutanoic acid
083015-50G
083015-10G
083015-1G
102838-670EA
767.66
USD
102838-670
102838-668
102838-666
4-Acetamidobutyric acid ≥97%
4-Acetamidobutyric acid
Hazard: Irritant
Formula:
C₆H₁₁NO₃ MW: 145.16 g/mol |
MDL Number:
MFCD00020552 CAS Number: 3025-96-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...