7-Bromoimidazo[1,2-a]pyridine-2-carboxylic acid ≥97%

Supplier: Matrix Scientific

082999-1G
102838-616EA 527.69 USD
102838-616
7-Bromoimidazo[1,2-a]pyridine-2-carboxylic acid ≥97%
7-Bromoimidazo[1,2-a]pyridine-2-carboxylic acid

Hazard: Irritant

Formula: C₈H₅BrN₂O₂
MDL Number: MFCD22989340
CAS Number: 1019018-46-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR