Imidazo[1,2-b]pyridazine-2-carboxylic acid ≥97%

Supplier: Matrix Scientific

082956-1G
102838-518EA 750.77 USD
102838-518
Imidazo[1,2-b]pyridazine-2-carboxylic acid ≥97%
Imidazo[1,2-b]pyridazine-2-carboxylic acid

Hazard: Irritant

Formula: C₇H₅N₃O₂
MW: 163.1300 g/mol
MDL Number: MFCD08235229
CAS Number: 160911-42-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR