trans-1,2-Cyclobutanedicarboxylic acid ≥97%
Supplier: Matrix Scientific
082878-10G
082878-1G
102838-326EA
284.62
USD
102838-326
102838-328
trans-1,2-Cyclobutanedicarboxylic acid ≥97%
trans-1,2-Cyclobutanedicarboxylic acid
Hazard: Irritant
Formula:
C₆H₈O₄ MW: 144.13 g/mol Melting Pt: 116…117 °C |
MDL Number:
MFCD00068260 CAS Number: 1124-13-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...