1-(3-Chlorophenyl)cyclopropanecarboxylic acid ≥97%

Supplier: Matrix Scientific

082868-1G
102838-298EA 509.23 USD
102838-298
1-(3-Chlorophenyl)cyclopropanecarboxylic acid ≥97%
1-(3-Chlorophenyl)cyclopropanecarboxylic acid

Hazard: Irritant

Formula: C₁₀H₉ClO₂
MW: 196.6300 g/mol
MDL Number: MFCD07374442
CAS Number: 124276-34-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR