4'-Methylbiphenyl-3-carboxylic acid ≥97%
Supplier: Matrix Scientific
Synonyms:
4'-Methyl-[1,1'-biphenyl]-3-carboxylic acid
082867-1G
102838-296EA
383.08
USD
102838-296
4'-Methylbiphenyl-3-carboxylic acid ≥97%
4'-Methylbiphenyl-3-carboxylic acid
Hazard: Irritant
Formula:
C₁₄H₁₂O₂ MW: 212.25 g/mol |
MDL Number:
MFCD00452725 CAS Number: 147404-69-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...