5-Bromo-2,3-dihydrobenzofuran-7-carboxylic acid ≥97%

Supplier: Matrix Scientific

Synonyms: 5-Bromocoumaran-7-carboxylic Acid

082721-1G
102837-976EA 213.85 USD
102837-976
5-Bromo-2,3-dihydrobenzofuran-7-carboxylic acid ≥97%
5-Bromo-2,3-dihydrobenzofuran-7-carboxylic acid

Hazard: Irritant

Formula: C₉H₇BrO₃
MW: 243.06 g/mol
MDL Number: MFCD00191391
CAS Number: 41177-72-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR