Benzo[b]thiophene-7-carboxylic acid ≥97%

Supplier: Matrix Scientific

082681-1G
102837-868EA 758.46 USD
102837-868
Benzo[b]thiophene-7-carboxylic acid ≥97%
Benzo[b]thiophene-7-carboxylic acid

Purity: 97%
CAS: 10134-98-2
MDL Number: MFCD08572208
Molecular Formula: C9H6O2S
Molecular Weight: 178,21
Hazard: Irritant

Formula: C₉H₆O₂S
CAS Number: 10134-98-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR