5-Oxazolecarboxylic acid ≥97%

Supplier: Matrix Scientific

Synonyms: Oxazole-5-carboxylic acid

082621-1G
102837-740EA 84.62 USD
102837-740
5-Oxazolecarboxylic acid ≥97%
5-Oxazolecarboxylic acid

Hazard: Irritant/Store in Freezer

Formula: C₄H₃NO₃
MW: 113.07 g/mol
Melting Pt: 243…247 °C
MDL Number: MFCD04114931
CAS Number: 118994-90-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR