5-(4-Chlorophenyl)-4-oxazolecarboxylic acid ≥97%

Supplier: Matrix Scientific

082605-1G
102837-708EA 602.6 USD
102837-708
5-(4-Chlorophenyl)-4-oxazolecarboxylic acid ≥97%
5-(4-Chlorophenyl)-4-oxazolecarboxylic acid

Hazard: Irritant

Formula: C₁₀H₆ClNO₃
MDL Number: MFCD00105450
CAS Number: 143659-14-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR