(±)-Tetrahydro-2-furoic acid ≥97%

Supplier: Matrix Scientific

Synonyms: 2-Tetrahydrofuran Carboxylic AcidTetrahydrofuran-2-carboxylic acid

082545-1G 082545-50G 082545-10G
102837-574EA 110.77 USD
102837-574 102837-572 102837-576
(±)-Tetrahydro-2-furoic acid ≥97%
(±)-Tetrahydro-2-furoic acid

Hazard: Corrosive

Formula: C₅H₈O₃
MW: 116.12 g/mol
Density: 1.209
MDL Number: MFCD00022473
CAS Number: 16874-33-2
UN: 3265
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR