2-(Phenylthio)nicotinic acid ≥97%

Supplier: Matrix Scientific

082469-1G
102837-402EA 353.85 USD
102837-402
2-(Phenylthio)nicotinic acid ≥97%
2-(Phenylthio)nicotinic acid

Hazard: Irritant

Formula: C₁₂H₉NO₂S
MW: 231.2704 g/mol
MDL Number: MFCD00052110
CAS Number: 35620-72-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR