2-(4-Fluorophenyl)isonicotinic acid ≥97%

Supplier: Matrix Scientific

082422-1G
102837-304EA 709.55 USD
102837-304
2-(4-Fluorophenyl)isonicotinic acid ≥97%
2-(4-Fluorophenyl)isonicotinic acid

Hazard: Irritant

Formula: C₁₂H₈FNO₂
MDL Number: MFCD14666481
CAS Number: 883528-25-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR