2-(4-Methylphenyl)isonicotinic acid ≥97%
Supplier: Matrix Scientific
082416-1G
102837-292EA
561.2
USD
102837-292
2-(4-Methylphenyl)isonicotinic acid ≥97%
2-(4-Methylphenyl)isonicotinic acid
Hazard: Irritant
Formula:
C₁₃H₁₁NO₂ |
MDL Number:
MFCD14666476 CAS Number: 1226205-64-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...