6-(3-Fluorophenyl)-2-pyridinecarboxylic acid ≥97%

Supplier: Matrix Scientific

082407-1G
102837-274EA 750.77 USD
102837-274
6-(3-Fluorophenyl)-2-pyridinecarboxylic acid ≥97%
6-(3-Fluorophenyl)-2-pyridinecarboxylic acid

Hazard: Irritant

Formula: C₁₂H₈FNO₂
MDL Number: MFCD06410053
CAS Number: 887982-40-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR