6-Amino-5-chloronicotinic acid ≥97%

Supplier: Matrix Scientific

082385-1G
102837-230EA 609.23 USD
102837-230
6-Amino-5-chloronicotinic acid ≥97%
6-Amino-5-chloronicotinic acid

Hazard: Irritant

Formula: C₆H₅ClN₂O₂
MDL Number: MFCD11132184
CAS Number: 41668-11-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR