2-(Cyanomethyl)benzoic acid ≥97%

Supplier: Matrix Scientific

082268-1G
102836-966EA 636.92 USD
102836-966
2-(Cyanomethyl)benzoic acid ≥97%
2-(Cyanomethyl)benzoic acid

Hazard: Irritant

Formula: C₉H₇NO₂
MW: 161.16 g/mol
CAS Number: 6627-91-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR