2,5-Dihydroxyterephthalic acid ≥97%

Supplier: Matrix Scientific

082241-1G
102836-898EA 253.69 USD
102836-898
2,5-Dihydroxyterephthalic acid ≥97%
2,5-Dihydroxyterephthalic acid

Purity: 97%
CAS: 610-92-4
MDL Number: MFCD00132933
Molecular Formula: C8H6O6
Molecular Weight: 198,13
Hazard: Irritant

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR