3,4-Bis(acetamido)benzoic acid ≥97%

Supplier: Matrix Scientific

Synonyms: 3,4-Diacetamidobenzoic acid

082227-1G
102836-862EA 241.54 USD
102836-862
3,4-Bis(acetamido)benzoic acid ≥97%
3,4-Bis(acetamido)benzoic acid

Hazard: Irritant

Formula: C₁₁H₁₂N₂O₄
MW: 236.23 g/mol
MDL Number: MFCD00238577
CAS Number: 205873-56-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR