2,6-Difluoro-4-hydroxybenzoic acid ≥97%

Supplier: Matrix Scientific

082184-1G
102836-762EA 164.62 USD
102836-762
2,6-Difluoro-4-hydroxybenzoic acid ≥97%
2,6-Difluoro-4-hydroxybenzoic acid

Hazard: Irritant

Formula: C₇H₄F₂O₃
MDL Number: MFCD03094500
CAS Number: 214917-68-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR