(S)-2-Benzyl-3-((tert-butoxycarbonyl)amino)propanoic acid ≥97%

Supplier: Matrix Scientific

Synonyms: (2S)-3-Amino-2-benzylpropanoic acid, N-BOC protected(S)-2-Benzyl-3-((tert-butoxycarbonyl)amino)propanoic acid(2S)-2-{[(tert-Butoxycarbonyl)amino]methyl}-3-phenylpropanoic acid

082129-1G
102836-636EA 1684.62 USD
102836-636
(S)-2-Benzyl-3-((tert-butoxycarbonyl)amino)propanoic acid ≥97%
(S)-2-Benzyl-3-((tert-butoxycarbonyl)amino)propanoic acid

Hazard: Irritant

Formula: C₁₅H₂₁NO₄
MW: 279.332 g/mol
MDL Number: MFCD07372495
CAS Number: 189619-55-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR