2-Ethylbenzoic acid ≥97%
Supplier: Matrix Scientific
082121-10G
082121-1G
102836-612EA
38.46
USD
102836-612
102836-614
2-Ethylbenzoic acid ≥97%
2-Ethylbenzoic acid
Hazard: Irritant
Formula:
C₉H₁₀O₂ MW: 150.1700 g/mol |
MDL Number:
MFCD00045838 CAS Number: 612-19-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...