4-(1H-1,2,4-Triazol-1-yl)benzoic acid ≥97%
Supplier: Matrix Scientific
Synonyms:
4-[1,2,4]Triazol-1-yl-benzoic acid
082076-10G
082076-1G
102836-490EA
298.46
USD
102836-490
102836-492
4-(1H-1,2,4-Triazol-1-yl)benzoic acid ≥97%
4-(1H-1,2,4-Triazol-1-yl)benzoic acid
Hazard: Irritant
Formula:
C₉H₇N₃O₂ MW: 189.1708 g/mol |
MDL Number:
MFCD02682060 CAS Number: 162848-16-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...