1-Phenylcyclobutanecarboxylic acid ≥97%

Supplier: Matrix Scientific

082053-1G 082053-10G
102836-428EA 307.69 USD
102836-428 102836-430
1-Phenylcyclobutanecarboxylic acid ≥97%
1-Phenylcyclobutanecarboxylic acid

Hazard: Irritant

Formula: C₁₀H₁₀O₂
MW: 176.22 g/mol
Melting Pt: 100…102 °C
MDL Number: MFCD00019262
CAS Number: 37828-19-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR