2-Acetylbenzoic acid ≥97%

Supplier: Matrix Scientific

082045-10G 082045-1G
102836-412EA 253.46 USD
102836-412 102836-410
2-Acetylbenzoic acid ≥97%
2-Acetylbenzoic acid

Hazard: Irritant

Formula: C₉H₈O₃
MW: 164.16 g/mol
MDL Number: MFCD00002475
CAS Number: 577-56-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR