2-Chloro-4-hydroxybenzoic acid hydrate ≥97%

Supplier: Matrix Scientific

Synonyms: 4-Carboxy-3-chlorophenol hydrate

082038-1G
102836-396EA 44.62 USD
102836-396
2-Chloro-4-hydroxybenzoic acid hydrate ≥97%
2-Chloro-4-hydroxybenzoic acid hydrate

Hazard: Irritant

Formula: C₇H₅ClO₃·xH₂O
MW: 172.57 g/mol (anhy)
MDL Number: MFCD00798116
CAS Number: 56363-84-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR