3-(1H-Pyrazol-3-yl)benzoic acid ≥97%

Supplier: Matrix Scientific

082033-1G
102836-378EA 308.2 USD
102836-378
3-(1H-Pyrazol-3-yl)benzoic acid ≥97%
3-(1H-Pyrazol-3-yl)benzoic acid

Hazard: Irritant

Formula: C₁₀H₈N₂O₂
MDL Number: MFCD03407956
CAS Number: 850375-11-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR