4-(4-Acetylpiperazin-1-yl)benzoic acid ≥97%

Supplier: Matrix Scientific

082022-1G
102836-350EA 453.85 USD
102836-350
4-(4-Acetylpiperazin-1-yl)benzoic acid ≥97%
4-(4-Acetylpiperazin-1-yl)benzoic acid

Hazard: Irritant

Formula: C₁₃H₁₆N₂O₃
MDL Number: MFCD05862631
CAS Number: 104080-55-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR