2-p-Terphenylboronic acid ≥97%

Supplier: Matrix Scientific

082009-1G
102836-318EA 509.23 USD
102836-318
2-p-Terphenylboronic acid ≥97%
2-p-Terphenylboronic acid

Hazard: Irritant

Formula: C₁₈H₁₅BO₂
CAS Number: 663954-31-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR