1-Ethyl-1H-pyrazole-4-boronic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 1-Ethylpyrazole-4-boronic Acid

082007-1G
102836-314EA 512.31 USD
102836-314
1-Ethyl-1H-pyrazole-4-boronic acid ≥95%
1-Ethyl-1H-pyrazole-4-boronic acid

Hazard: Irritant

Formula: C₅H₉BN₂O₂
MW: 139.9500 g/mol
MDL Number: MFCD10703521
CAS Number: 847818-56-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR