3-(Morpholinomethyl)phenylboronic acid pinacol ester ≥97%

Supplier: Matrix Scientific

Synonyms: 4-(3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl)morpholine3-(4-Morpholinylmethyl)benzeneboronic acid pinacol ester

081981-1G
102836-256EA 467.69 USD
102836-256
3-(Morpholinomethyl)phenylboronic acid pinacol ester ≥97%
3-(Morpholinomethyl)phenylboronic acid pinacol ester

Hazard: Irritant

Formula: C₁₇H₂₆BNO₃
MW: 303.20 g/mol
MDL Number: MFCD06797489
CAS Number: 364794-80-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR