4-(Morpholinomethyl)phenylboronic acid pinacol ester ≥97%

Supplier: Matrix Scientific

Synonyms: 4-[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl]morpholine4-(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl)morpholine4,4,5,5-Tetramethyl-2-[4-(morpholinomethyl)phenyl]-1,3,2-dioxaborolane4-(4-Morpholinylmethyl)benzeneboronic acid pinacol ester4-(Morpholinomethyl)phenylboronic acid Pinacol ester

081976-1G
102836-246EA 172.31 USD
102836-246
4-(Morpholinomethyl)phenylboronic acid pinacol ester ≥97%
4-(Morpholinomethyl)phenylboronic acid pinacol ester

Hazard: Irritant

Formula: C₁₇H₂₆BNO₃
MW: 303.20 g/mol
MDL Number: MFCD04974052
CAS Number: 364794-79-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR