N-(2,2-Diphenylethyl)-N-methylamine ≥97%

Supplier: Matrix Scientific

081967-1G
102836-230EA 524.62 USD
102836-230
N-(2,2-Diphenylethyl)-N-methylamine ≥97%
N-(2,2-Diphenylethyl)-N-methylamine

Hazard: Irritant

Formula: C₁₅H₁₇N
MW: 211.31 g/mol
MDL Number: MFCD14666467
CAS Number: 80376-82-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR