N-Methyl-N-(2-pyridin-3-ylethyl)amine ≥97%

Supplier: Matrix Scientific

Synonyms: N-Methyl-2-(pyridin-3-yl)ethanamine

081883-1G
102836-038EA 863.08 USD
102836-038
N-Methyl-N-(2-pyridin-3-ylethyl)amine ≥97%
N-Methyl-N-(2-pyridin-3-ylethyl)amine

Hazard: Irritant

Formula: C₈H₁₂N₂
MW: 136.2 g/mol
MDL Number: MFCD02089408
CAS Number: 19690-13-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR