N-[2-(3-Chlorophenyl)ethyl]-N-methylamine ≥97%

Supplier: Matrix Scientific

081843-1G
102835-954EA 949.23 USD
102835-954
N-[2-(3-Chlorophenyl)ethyl]-N-methylamine ≥97%
N-[2-(3-Chlorophenyl)ethyl]-N-methylamine

Hazard: Irritant

Formula: C₉H₁₂ClN
MDL Number: MFCD06738721
CAS Number: 52516-20-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR