1-(4-Chlorophenyl)cyclopropanamine ≥97%

Supplier: Matrix Scientific

081823-1G
102835-908EA 949.23 USD
102835-908
1-(4-Chlorophenyl)cyclopropanamine ≥97%
1-(4-Chlorophenyl)cyclopropanamine

Hazard: Irritant

Formula: C₉H₁₀ClN
MW: 167.64 g/mol
MDL Number: MFCD07371527
CAS Number: 72934-36-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR