9-Julolidinecarboxaldehyde ≥97%

Supplier: Matrix Scientific

Synonyms: 2,3,6,7-Tetrahydro-1H,5H-benzo[ij]quinolizine-9-carboxaldehyde1,2,3,5,6,7-Hexahydropyrido[3,2,1-ij]quinoline-9-carbaldehyde

081750-1G
102835-728EA 553.85 USD
102835-728
9-Julolidinecarboxaldehyde ≥97%
9-Julolidinecarboxaldehyde

Hazard: Irritant

Formula: C₁₃H₁₅NO
MW: 201.27 g/mol
MDL Number: MFCD00151555
CAS Number: 33985-71-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR