1-Naphthaldehyde ≥97%

Supplier: Matrix Scientific

Synonyms: α-Naphthaldehyde

081747-1G 081747-10G
102835-718EA 44.62 USD
102835-718 102835-720
1-Naphthaldehyde ≥97%
1-Naphthaldehyde

Hazard: Irritant

Formula: C₁₁H₈O
MW: 156.18 g/mol
MDL Number: MFCD00004003
CAS Number: 66-77-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR