6,7-Dihydro-5H-pyrrolo[1,2-a]imidazole-3-carbaldehyde ≥95%

Supplier: Matrix Scientific

081741-1G
102835-704EA 806.15 USD
102835-704
6,7-Dihydro-5H-pyrrolo[1,2-a]imidazole-3-carbaldehyde ≥95%
6,7-Dihydro-5H-pyrrolo[1,2-a]imidazole-3-carbaldehyde

Hazard: Irritant

Formula: C₇H₈N₂O
MW: 136.15 g/mol
MDL Number: MFCD08235198
CAS Number: 914637-04-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR