Cyclopropanecarboxaldehyde ≥97%

Supplier: Matrix Scientific

Synonyms: Cyclopropanecarbaldehyde

081739-5G 081739-25G
102835-700EA 112.31 USD
102835-700 102835-698
Cyclopropanecarboxaldehyde ≥97%
Cyclopropanecarboxaldehyde

Purity: 97%
CAS: 1489-69-6
MDL Number: MFCD00012261
Molecular Formula: C4H6O
Molecular Weight: 70,09
Boiling Point: 98-101°C
Hazard: Flammable/Corrosive/Refrigerate

Formula: C₄H₆O
MW: 70.09 g/mol
Boiling Pt: 98…101 °C
Density: 0.938
MDL Number: MFCD00012261
CAS Number: 1489-69-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR