Diphenylacetaldehyde ≥97%

Supplier: Matrix Scientific

Synonyms: 2,2-Diphenylacetaldehyde

081738-1G 081738-10G
102835-694EA 107.69 USD
102835-694 102835-696
Diphenylacetaldehyde ≥97%
Diphenylacetaldehyde

Hazard: Irritant

Formula: C₁₄H₁₂O
MW: 196.2400 g/mol
Boiling Pt: 315 °C
Density: 1.106
Flash Pt: 113 °C
MDL Number: MFCD00006972
CAS Number: 947-91-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR